Product Details

SNP ID
rs171407
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.3:10284485 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GATTCTAAGCCATTTCCCATCAGCC[A/G]TTTACTAAGTAGCCTGGGGATCTTG
Phenotype
MIM: 605353
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
GHRL PubMed Links

Gene Details

Gene
GHRL
Gene Name
ghrelin/obestatin prepropeptide
There are no transcripts associated with this gene.

Gene
GHRLOS
Gene Name
ghrelin opposite strand/antisense RNA
There are no transcripts associated with this gene.

Gene
LINC00852
Gene Name
long intergenic non-protein coding RNA 852
There are no transcripts associated with this gene.

Gene
TATDN2
Gene Name
TatD DNase domain containing 2
There are no transcripts associated with this gene.

View Full Product Details