Product Details

SNP ID
rs1640702
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:10976987 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGAAAATGATTGTATTTAAAATGAT[C/T]AGTAGTCAACAGAAATTGAATCATA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PHF14 PubMed Links

Gene Details

Gene
PHF14
Gene Name
PHD finger protein 14
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014660.3 Intron NP_055475.2
XM_017012811.1 Intron XP_016868300.1
XM_017012812.1 Intron XP_016868301.1
XM_017012813.1 Intron XP_016868302.1
XM_017012814.1 Intron XP_016868303.1
XM_017012815.1 Intron XP_016868304.1
XM_017012816.1 Intron XP_016868305.1
XM_017012817.1 Intron XP_016868306.1

View Full Product Details