Product Details

SNP ID
rs3784310
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:71811087 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCTGCAGAGCCAGGACAGGACAGCC[C/T]AGCCGATGGGGAAGGAAAGAACAGA
Phenotype
MIM: 604485
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NR2E3 PubMed Links

Gene Details

Gene
NR2E3
Gene Name
nuclear receptor subfamily 2 group E member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014249.3 Intron NP_055064.1
NM_016346.3 Intron NP_057430.1

View Full Product Details