Product Details

SNP ID
rs684227
Assay Type
Functionally tested
NCBI dbSNP Submissions
50
Location
Chr.1:177932606 on Build GRCh38
Set Membership
HapMap JSNP
Context Sequence [VIC/FAM]
TGAGGCAGAGCTGTTTCCTCTGCTC[A/G]GGACTGGGGGTCCCCACTCCCAGAA
Phenotype
MIM: 612855
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SEC16B PubMed Links

Gene Details

Gene
SEC16B
Gene Name
SEC16 homolog B, endoplasmic reticulum export factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033127.2 Intron NP_149118.2

View Full Product Details