Product Details

SNP ID
rs7689420
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:144647200 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACTTGCCTATCTCTGAAAACAGACG[C/T]TTCTGCGGTACCCGCACCCTGGGCT
Phenotype
MIM: 606178
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HHIP PubMed Links

Gene Details

Gene
HHIP
Gene Name
hedgehog interacting protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022475.2 Intron NP_071920.1
XM_005263178.4 Intron XP_005263235.1
XM_006714288.3 Intron XP_006714351.1
Gene
HHIP-AS1
Gene Name
HHIP antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details