Product Details

SNP ID
rs1047720
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:14588912 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GACTCAGATGATCAAGCCGATATCA[A/G]TGAGGAAAATGGAGTACTATCCCTC
Phenotype
MIM: 614605
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZDHHC21 PubMed Links

Gene Details

Gene
ZDHHC21
Gene Name
zinc finger DHHC-type containing 21
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178566.4 Intron NP_848661.1
XM_017014682.1 Intron XP_016870171.1
XM_017014683.1 Intron XP_016870172.1
XM_017014684.1 Intron XP_016870173.1
XM_017014685.1 Intron XP_016870174.1
XM_017014686.1 Intron XP_016870175.1
XM_017014687.1 Intron XP_016870176.1
XM_017014688.1 Intron XP_016870177.1
XM_017014689.1 Intron XP_016870178.1
XM_017014690.1 Intron XP_016870179.1
XM_017014691.1 Intron XP_016870180.1
XM_017014692.1 Intron XP_016870181.1
XM_017014693.1 Intron XP_016870182.1

View Full Product Details