Product Details

SNP ID
rs13463
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.12:51095226 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
TGAAGGAAGGAGCAGCCACTGGGCA[C/T]GAAACGCCGCACTCCTACTTCAGTA
Phenotype
MIM: 189889
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TFCP2 PubMed Links

Gene Details

Gene
TFCP2
Gene Name
transcription factor CP2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001173452.1 2245 UTR 3 NP_001166923.1
NM_001173453.1 2245 UTR 3 NP_001166924.1
NM_005653.4 2245 UTR 3 NP_005644.2

View Full Product Details