Product Details

SNP ID
rs907091
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:39765489 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAGAATGTTTCATATAGCACATCTC[C/T]GGGACCACTCATTCCACTGCTGTAG
Phenotype
MIM: 606221
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IKZF3 PubMed Links

Gene Details

Gene
IKZF3
Gene Name
IKAROS family zinc finger 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001257408.1 1674 UTR 3 NP_001244337.1
NM_001257409.1 1674 UTR 3 NP_001244338.1
NM_001257410.1 1674 UTR 3 NP_001244339.1
NM_001257411.1 1674 UTR 3 NP_001244340.1
NM_001257412.1 1674 UTR 3 NP_001244341.1
NM_001257413.1 1674 UTR 3 NP_001244342.1
NM_001257414.1 1674 UTR 3 NP_001244343.1
NM_001284514.1 1674 UTR 3 NP_001271443.1
NM_001284515.1 1674 UTR 3 NP_001271444.1
NM_001284516.1 1674 UTR 3 NP_001271445.1
NM_012481.4 1674 UTR 3 NP_036613.2
NM_183228.2 1674 UTR 3 NP_899051.1
NM_183229.2 1674 UTR 3 NP_899052.1
NM_183230.2 1674 UTR 3 NP_899053.1
NM_183231.2 1674 UTR 3 NP_899054.1
NM_183232.2 1674 UTR 3 NP_899055.1

View Full Product Details