Product Details

SNP ID
rs1781933
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:5156988 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAAGGTCCAGAGTGGATGTGGAGAA[A/G]GGTTTACAAGATAACTTCACTCCTC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
AKR1C8P PubMed Links

Gene Details

Gene
AKR1C8P
Gene Name
aldo-keto reductase family 1 member C8, pseudogene
There are no transcripts associated with this gene.

View Full Product Details