Product Details

SNP ID
rs998544
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:56417412 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GATCACCACTGTGTAAAATACAGAA[A/G]CCATCTTGTCTGTGTCCAAGGAGTG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR5R1 PubMed Links

Gene Details

Gene
OR5R1
Gene Name
olfactory receptor family 5 subfamily R member 1 (gene/pseudogene)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004744.1 821 Missense Mutation GCT,GTT A274V NP_001004744.1

View Full Product Details