Product Details
- SNP ID
-
rs868641
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:151105680 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CAGGGTTTAGGCTCCACCCTAAAGA[C/T]TCAGATTTTGGAAAGATTTTGAGCA
- Phenotype
-
MIM: 114070
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ANXA6
PubMed Links
Gene Details
- Gene
- ANXA6
- Gene Name
- annexin A6
View Full Product Details