Product Details

SNP ID
rs1050381
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:43224786 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCCCCGGCGCCAGCAGGTCCTCGG[A/C]GCCCCCGGTGCCGTACAGCAGAGCG
Phenotype
MIM: 611800 MIM: 612053
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
LINC01126 PubMed Links

Gene Details

Gene
LINC01126
Gene Name
long intergenic non-protein coding RNA 1126
There are no transcripts associated with this gene.

Gene
THADA
Gene Name
THADA, armadillo repeat containing
There are no transcripts associated with this gene.

Gene
ZFP36L2
Gene Name
ZFP36 ring finger protein like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006887.4 1309 Missense Mutation GCC,TCC A340S NP_008818.3

View Full Product Details