Product Details
- SNP ID
-
rs8698
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
42
- Location
-
Chr.1:236549557 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTTTAAAAGTACTCAGTGTAGAAAT[C/T]GCTAGCCCTTAATTCTTTTCCAGCT
- Phenotype
-
MIM: 606099
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
HEATR1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs76792063] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- HEATR1
- Gene Name
- HEAT repeat containing 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_018072.5 |
2731 |
UTR 3 |
|
|
NP_060542.4 |
- Gene
- LGALS8
- Gene Name
- galectin 8
View Full Product Details