Product Details

SNP ID
rs889300
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:95856513 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGAAAGAAACAGCATTGTGTGGTGT[A/C]CCCCAGCACCTCCCAGACAAAAGGC
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
LINC01554 PubMed Links

Gene Details

Gene
LINC01554
Gene Name
long intergenic non-protein coding RNA 1554
There are no transcripts associated with this gene.

Gene
LOC102724720
Gene Name
uncharacterized LOC102724720
There are no transcripts associated with this gene.

View Full Product Details