Product Details
- SNP ID
-
rs860290
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:5453198 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AGAAATAAAAAAATAGGTACCTAAC[A/C]CTGGCAAGAGGATGATGACTCATGT
- Phenotype
-
MIM: 605402
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
CD274
PubMed Links
Gene Details
- Gene
- CD274
- Gene Name
- CD274 molecule
View Full Product Details