Product Details

SNP ID
rs944626
Assay Type
Functionally tested
NCBI dbSNP Submissions
14
Location
Chr.1:157677578 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGTCATATTAAAAATTCAACACACT[C/G]TGGAAAGAGTTTTTGATGGTGATAG
Phenotype
MIM: 606510
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
FCRL3 PubMed Links

Gene Details

Gene
FCRL3
Gene Name
Fc receptor like 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320333.1 3559 Intron NP_001307262.1
NM_052939.3 3559 Intron NP_443171.2
XM_006711145.1 3559 UTR 3 XP_006711208.1

View Full Product Details