Product Details

SNP ID
rs1708085
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:39468159 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGTTTTTCTTCTTATCTAGGAACAA[C/T]TTTTTTCCTCATACAGTTGGATTTA
Phenotype
MIM: 600948
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MOBP PubMed Links

Gene Details

Gene
MOBP
Gene Name
myelin-associated oligodendrocyte basic protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278322.1 Intron NP_001265251.1
NM_001278323.1 Intron NP_001265252.1
NM_182935.3 Intron NP_891980.1

View Full Product Details