Product Details

SNP ID
rs954535
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:225488908 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTGCCAAGACATTGCTGTTAAACA[A/G]AGTCTTTTGGATTAAAACCAGGGAA
Phenotype
MIM: 609061
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ENAH PubMed Links

Gene Details

Gene
ENAH
Gene Name
enabled homolog (Drosophila)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001008493.1 10862 UTR 3 NP_001008493.1
NM_018212.4 10862 UTR 3 NP_060682.2
XM_005273187.2 10862 UTR 3 XP_005273244.1
XM_011544229.2 10862 Intron XP_011542531.2
XM_017001746.1 10862 UTR 3 XP_016857235.1
XM_017001747.1 10862 Intron XP_016857236.1
XM_017001748.1 10862 Intron XP_016857237.1
XM_017001749.1 10862 Intron XP_016857238.1
XM_017001750.1 10862 Intron XP_016857239.1
XM_017001751.1 10862 UTR 3 XP_016857240.1
XM_017001752.1 10862 Intron XP_016857241.1
XM_017001753.1 10862 UTR 3 XP_016857242.1
XM_017001754.1 10862 UTR 3 XP_016857243.1
XM_017001755.1 10862 UTR 3 XP_016857244.1
XM_017001756.1 10862 UTR 3 XP_016857245.1
XM_017001757.1 10862 UTR 3 XP_016857246.1

View Full Product Details