Product Details

SNP ID
rs1062826
Assay Type
Validated
NCBI dbSNP Submissions
47
Location
Chr.1:160998215 on Build GRCh38
Set Membership
HapMap JSNP Validated
Context Sequence [VIC/FAM]
TAGAGCAGCGGTTTTAAATCCTTTT[C/G]GCTTCAAGTTCTCTGAAAATTTACT
Phenotype
MIM: 605721
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
F11R PubMed Links

Gene Details

Gene
F11R
Gene Name
F11 receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016946.4 1826 UTR 3 NP_058642.1

View Full Product Details