Product Details
- SNP ID
-
rs6715929
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:172012600 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- AAATCACAGCTGCGGAAGACAGTCT[C/T]GCAAAGCCAGTTTTTGTGGCTTAGT
- Phenotype
-
MIM: 610267
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
METAP1D
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs62183807] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- METAP1D
- Gene Name
- methionyl aminopeptidase type 1D (mitochondrial)
View Full Product Details