Product Details

SNP ID
rs10944288
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.6:86942024 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
ATGCCATTAGAAAGAATAAGAGAAC[C/T]CAGTAAGGTGGCTAGCTACAATAGC
Phenotype
MIM: 182132
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HTR1E PubMed Links

Gene Details

Gene
HTR1E
Gene Name
5-hydroxytryptamine receptor 1E
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000865.2 Intron NP_000856.1
XM_011535789.2 Intron XP_011534091.1

View Full Product Details