Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_015836.3 | 1297 | UTR 3 | NP_056651.1 | ||
NM_201263.2 | 1297 | UTR 3 | NP_957715.1 | ||
XM_005270350.3 | 1297 | UTR 3 | XP_005270407.1 | ||
XM_011540494.2 | 1297 | UTR 3 | XP_011538796.1 | ||
XM_011540495.2 | 1297 | UTR 3 | XP_011538797.1 | ||
XM_017000038.1 | 1297 | UTR 3 | XP_016855527.1 | ||
XM_017000039.1 | 1297 | UTR 3 | XP_016855528.1 | ||
XM_017000040.1 | 1297 | UTR 3 | XP_016855529.1 | ||
XM_017000041.1 | 1297 | UTR 3 | XP_016855530.1 | ||
XM_017000042.1 | 1297 | UTR 3 | XP_016855531.1 |