Product Details

SNP ID
rs3850817
Assay Type
Functionally tested
NCBI dbSNP Submissions
28
Location
Chr.1:247143230 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCAGGACATGCCTTCAAGACACACC[C/G]AAATGCCTATATATATAATAATACT
Phenotype
MIM: 194631
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ZNF124 PubMed Links

Gene Details

Gene
ZNF124
Gene Name
zinc finger protein 124
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001243740.2 Intron NP_001230669.1
NM_001297567.1 Intron NP_001284496.1
NM_001297568.1 Intron NP_001284497.1
NM_001297569.1 Intron NP_001284498.1
NM_003431.3 Intron NP_003422.2
XM_005273257.4 Intron XP_005273314.1
XM_006711813.3 Intron XP_006711876.1
XM_011544272.2 Intron XP_011542574.1

View Full Product Details