Product Details

SNP ID
rs729636
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.17:41812449 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CATCCCAAGTCACAAAGTCCTTCTT[G/T]CAGTCCAGCTTTTGCGGGACCAAGC
Phenotype
MIM: 607063
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
FKBP10 PubMed Links

Gene Details

Gene
FKBP10
Gene Name
FK506 binding protein 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021939.3 188 Intron NP_068758.3
XM_011525099.2 188 UTR 5 XP_011523401.1
XM_011525100.2 188 Intron XP_011523402.1
Gene
P3H4
Gene Name
prolyl 3-hydroxylase family member 4 (non-enzymatic)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006455.2 188 Intron NP_006446.1
XM_006721640.3 188 UTR 5 XP_006721703.1

View Full Product Details