Product Details

SNP ID
rs1056545
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:55430730 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCTTGAAATTATTATATTCTAAAAT[A/G]TTACTAGCAATCACTTTTAGATTAG
Phenotype
MIM: 601851 MIM: 614726
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CLOCK PubMed Links

Gene Details

Gene
CLOCK
Gene Name
clock circadian regulator
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001267843.1 7238 UTR 3 NP_001254772.1
NM_004898.3 7238 UTR 3 NP_004889.1
XM_005265787.2 7238 UTR 3 XP_005265844.1
XM_011534409.2 7238 UTR 3 XP_011532711.1
XM_011534410.2 7238 UTR 3 XP_011532712.1
XM_011534411.2 7238 UTR 3 XP_011532713.1
XM_017008854.1 7238 UTR 3 XP_016864343.1
XM_017008855.1 7238 UTR 3 XP_016864344.1
Gene
TMEM165
Gene Name
transmembrane protein 165
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018475.4 7238 Intron NP_060945.2
XM_011534394.2 7238 Intron XP_011532696.1
XM_017008412.1 7238 Intron XP_016863901.1

View Full Product Details