Product Details

SNP ID
rs1057631
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:40712142 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATTCATTTCACATTCTTCCCTTCCA[A/T]TTCAATCTAGAGTCAACAATGTGGT
Phenotype
MIM: 601586
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
PTGER4 PubMed Links

Gene Details

Gene
PTGER4
Gene Name
prostaglandin E receptor 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000958.2 4940 Intron NP_000949.1
XM_017009656.1 4940 Intron XP_016865145.1
XM_017009657.1 4940 Intron XP_016865146.1
XM_017009658.1 4940 Intron XP_016865147.1
XM_017009659.1 4940 Intron XP_016865148.1
Gene
TTC33
Gene Name
tetratricopeptide repeat domain 33
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012382.2 4940 UTR 3 NP_036514.1
XM_011514003.2 4940 Intron XP_011512305.1

View Full Product Details