Product Details

SNP ID
rs1053448
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:32775733 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGATTTCTCCAGCTGCCAAGGGAA[A/G]AAGGTAGGGCTGGACTCCCTGCTGT
Phenotype
MIM: 603623
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KIAA1522 PubMed Links

Gene Details

Gene
KIAA1522
Gene Name
KIAA1522
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001198972.1 1622 Intron NP_001185901.1
NM_001198973.1 1622 Intron NP_001185902.1
NM_020888.2 1622 Intron NP_065939.2
XM_006710791.3 1622 Intron XP_006710854.1
XM_017001916.1 1622 Intron XP_016857405.1
XM_017001917.1 1622 Intron XP_016857406.1
Gene
YARS
Gene Name
tyrosyl-tRNA synthetase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003680.3 1622 UTR 3 NP_003671.1
XM_011542347.1 1622 UTR 3 XP_011540649.1
XM_011542348.1 1622 UTR 3 XP_011540650.1
XM_017002651.1 1622 UTR 3 XP_016858140.1

View Full Product Details