Product Details
- SNP ID
-
rs3859315
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.18:79682583 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CGAGGGCCACCTTTATGTAGATGTT[G/T]TTCAGTAAATCGTCTGCTGTTCGCA
- Phenotype
-
MIM: 604927
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
CTDP1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs74337849] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CTDP1
- Gene Name
- CTD phosphatase subunit 1
- Gene
- LOC105376875
- Gene Name
- uncharacterized LOC105376875
- Gene
- LOC284241
- Gene Name
- uncharacterized LOC284241
There are no transcripts associated with this gene.
View Full Product Details