Product Details
- SNP ID
-
rs1767031
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
37
- Location
-
Chr.1:108820200 on Build GRCh38
- Set Membership
-
HapMap
JSNP
- Context Sequence [VIC/FAM]
- TGCTAGTTCGTTCATACTTATCCTT[C/T]GAAGCTCAGCTTCCCTGAGCCCCCA
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
AKNAD1
PubMed Links
Gene Details
- Gene
- AKNAD1
- Gene Name
- AKNA domain containing 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_152763.4 |
|
Intron |
|
|
NP_689976.2 |
View Full Product Details