Product Details

SNP ID
rs1548620
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:39750148 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAACAGACTCACCTGTTGGTATCTT[C/G]TTCATTTTTTTCAAAGCCAAACCAG
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LINC00265 PubMed Links

Gene Details

Gene
LINC00265
Gene Name
long intergenic non-protein coding RNA 265
There are no transcripts associated with this gene.

View Full Product Details