Product Details

SNP ID
rs1063816
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:241594504 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTGTCGTCAACTGACAGTGATTCA[C/T]CACTGGTGATGATAAAAATGATGGA
Phenotype
MIM: 603538 MIM: 606695
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KMO PubMed Links

Gene Details

Gene
KMO
Gene Name
kynurenine 3-monooxygenase (kynurenine 3-hydroxylase)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003679.4 1278 UTR 3 NP_003670.2
XM_005273337.1 1278 Intron XP_005273394.1
XM_005273338.3 1278 Intron XP_005273395.1
XM_017002649.1 1278 Intron XP_016858138.1
XM_017002650.1 1278 Intron XP_016858139.1
Gene
OPN3
Gene Name
opsin 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014322.2 1278 Missense Mutation GAT,GGT D378G NP_055137.2

View Full Product Details