Product Details

SNP ID
rs748113
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:71749034 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAGGAAGTGCGGTGGCTGTTCTGC[C/T]GCGGGCCGCCTCCCTCTCCACACAG
Phenotype
MIM: 615608 MIM: 605516
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C10orf54 PubMed Links

Gene Details

Gene
C10orf54
Gene Name
chromosome 10 open reading frame 54
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022153.1 3296 UTR 3 NP_071436.1
Gene
CDH23
Gene Name
cadherin-related 23
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001171930.1 3296 Intron NP_001165401.1
NM_001171931.1 3296 Intron NP_001165402.1
NM_001171932.1 3296 Intron NP_001165403.1
NM_001171933.1 3296 Intron NP_001165404.1
NM_001171934.1 3296 Intron NP_001165405.1
NM_001171935.1 3296 Intron NP_001165406.1
NM_001171936.1 3296 Intron NP_001165407.1
NM_022124.5 3296 Intron NP_071407.4
NM_052836.3 3296 Intron NP_443068.1
XM_006717940.3 3296 Intron XP_006718003.1
XM_006717942.3 3296 Intron XP_006718005.1
XM_011540039.2 3296 Intron XP_011538341.1
XM_011540042.2 3296 Intron XP_011538344.1
XM_011540043.2 3296 Intron XP_011538345.1
XM_011540044.2 3296 Intron XP_011538346.1
XM_011540045.2 3296 Intron XP_011538347.1
XM_011540046.2 3296 Intron XP_011538348.1
XM_011540047.2 3296 Intron XP_011538349.1
XM_011540048.2 3296 Intron XP_011538350.1
XM_011540049.2 3296 Intron XP_011538351.1
XM_011540051.2 3296 Intron XP_011538353.1
XM_011540052.2 3296 Intron XP_011538354.1
XM_017016499.1 3296 Intron XP_016871988.1
XM_017016500.1 3296 Intron XP_016871989.1
XM_017016501.1 3296 Intron XP_016871990.1
XM_017016502.1 3296 Intron XP_016871991.1
XM_017016503.1 3296 Intron XP_016871992.1
XM_017016504.1 3296 Intron XP_016871993.1
XM_017016505.1 3296 Intron XP_016871994.1
XM_017016506.1 3296 Intron XP_016871995.1
XM_017016507.1 3296 Intron XP_016871996.1
XM_017016508.1 3296 Intron XP_016871997.1

View Full Product Details