Product Details

SNP ID
rs2007257
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:54517868 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTCGGGGCTCCAGTCCGCGCGCCA[A/G]GGCTCGAGCAGTACCGCGGGCCCCT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SNX18 PubMed Links

Gene Details

Gene
SNX18
Gene Name
sorting nexin 18
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001102575.1 106 UTR 5 NP_001096045.1
NM_001145427.1 106 UTR 5 NP_001138899.1
NM_052870.2 106 UTR 5 NP_443102.2
XM_017008997.1 106 UTR 5 XP_016864486.1

View Full Product Details