Product Details

SNP ID
rs1047792
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:6539933 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTCCACCTGGCAGGTGGACAGCGT[G/T]AGCACGTGGACCATAGCAGGGACAA
Phenotype
MIM: 138400 MIM: 610495 MIM: 615638
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
GAPDH PubMed Links

Gene Details

Gene
GAPDH
Gene Name
glyceraldehyde-3-phosphate dehydrogenase
There are no transcripts associated with this gene.

Gene
IFFO1
Gene Name
intermediate filament family orphan 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039670.2 2579 UTR 3 NP_001034759.1
NM_001193457.1 2579 UTR 3 NP_001180386.1
NM_080730.4 2579 UTR 3 NP_542768.2
XM_006718975.3 2579 UTR 3 XP_006719038.1
XM_006718976.3 2579 UTR 3 XP_006719039.1
XM_006718977.3 2579 UTR 3 XP_006719040.1
XM_006718981.3 2579 UTR 3 XP_006719044.1
XM_011520947.2 2579 UTR 3 XP_011519249.1
XM_011520948.2 2579 UTR 3 XP_011519250.1
XM_011520949.2 2579 UTR 3 XP_011519251.1
XM_011520952.2 2579 UTR 3 XP_011519254.1
XM_017019169.1 2579 Intron XP_016874658.1
XM_017019170.1 2579 UTR 3 XP_016874659.1
XM_017019171.1 2579 UTR 3 XP_016874660.1
XM_017019172.1 2579 UTR 3 XP_016874661.1
XM_017019173.1 2579 UTR 3 XP_016874662.1
Gene
NCAPD2
Gene Name
non-SMC condensin I complex subunit D2
There are no transcripts associated with this gene.

View Full Product Details