Product Details
- SNP ID
-
rs1134597
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:3539110 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- GGACTTCTGATGGGAAATCTGAACT[A/G]CTGTTTCTCTTTTAGCGTACAAGAA
- Phenotype
-
MIM: 616787
MIM: 615648
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CLUAP1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs74005831] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CLUAP1
- Gene Name
- clusterin associated protein 1
- Gene
- LOC101929732
- Gene Name
- uncharacterized LOC101929732
There are no transcripts associated with this gene.
- Gene
- NLRC3
- Gene Name
- NLR family CARD domain containing 3
View Full Product Details