Product Details

SNP ID
rs894369
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:99412689 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAACTGATTTCAAAATGTAATCCAA[C/G]TTAACTCATAACAAGTGTTTAAAAA
Phenotype
MIM: 600086
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ADH7 PubMed Links

Gene Details

Gene
ADH7
Gene Name
alcohol dehydrogenase 7 (class IV), mu or sigma polypeptide
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000673.4 1861 UTR 3 NP_000664.2
NM_001166504.1 1861 UTR 3 NP_001159976.1

View Full Product Details