Product Details

SNP ID
rs9427662
Assay Type
Validated
NCBI dbSNP Submissions
58
Location
Chr.1:196977645 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CATCACTGGAGTATTTTTAGTTATA[C/T]ACGATTGAGACTACCAAGCATGTTG
Phenotype
MIM: 608593
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CFHR5 PubMed Links

Gene Details

Gene
CFHR5
Gene Name
complement factor H related 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030787.3 109 UTR 5 NP_110414.1
XM_011510020.2 109 Intron XP_011508322.1

View Full Product Details