Product Details

SNP ID
rs1317672
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:124043983 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAATGTCTGGCCATCAGGGTACTGT[A/G]TCAGTGATGGGATTTGCTCATGAAT
Phenotype
MIM: 604605
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KALRN PubMed Links

Gene Details

Gene
KALRN
Gene Name
kalirin, RhoGEF kinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001024660.4 Intron NP_001019831.2
NM_001322988.1 Intron NP_001309917.1
NM_001322989.1 Intron NP_001309918.1
NM_001322990.1 Intron NP_001309919.1
NM_001322991.1 Intron NP_001309920.1
NM_001322992.1 Intron NP_001309921.1
NM_001322993.1 Intron NP_001309922.1
NM_001322994.1 Intron NP_001309923.1
NM_001322995.1 Intron NP_001309924.1
NM_001322996.1 Intron NP_001309925.1
NM_001322997.1 Intron NP_001309926.1
NM_001322998.1 Intron NP_001309927.1
NM_001322999.1 Intron NP_001309928.1
NM_001323000.1 Intron NP_001309929.1
NM_001323001.1 Intron NP_001309930.1
NM_003947.5 Intron NP_003938.1
NM_007064.4 Intron NP_008995.2
XM_006713810.3 Intron XP_006713873.1
XM_006713811.3 Intron XP_006713874.1
XM_006713812.3 Intron XP_006713875.1
XM_006713813.3 Intron XP_006713876.1
XM_006713814.3 Intron XP_006713877.1
XM_006713815.3 Intron XP_006713878.1
XM_011513279.2 Intron XP_011511581.1
XM_011513280.2 Intron XP_011511582.1
XM_011513281.2 Intron XP_011511583.1
XM_011513283.2 Intron XP_011511585.1
XM_011513285.2 Intron XP_011511587.1
XM_017007429.1 Intron XP_016862918.1
XM_017007430.1 Intron XP_016862919.1
XM_017007431.1 Intron XP_016862920.1
XM_017007432.1 Intron XP_016862921.1
XM_017007433.1 Intron XP_016862922.1
XM_017007434.1 Intron XP_016862923.1
XM_017007435.1 Intron XP_016862924.1

View Full Product Details