Product Details

SNP ID
rs1609696
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.16:1159177 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACGTCTTCATGGCCAGTTCCCCGC[A/G]GAGTGCCCGGGTGTCCGTATCCCGG
Phenotype
MIM: 607904
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
CACNA1H PubMed Links

Gene Details

Gene
CACNA1H
Gene Name
calcium voltage-gated channel subunit alpha1 H
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005407.1 Intron NP_001005407.1
NM_021098.2 Intron NP_066921.2
XM_005255652.4 Intron XP_005255709.1
XM_006720963.3 Intron XP_006721026.1
XM_006720964.3 Intron XP_006721027.1
XM_006720965.3 Intron XP_006721028.1
XM_006720967.3 Intron XP_006721030.1
XM_006720968.3 Intron XP_006721031.1
XM_011522724.2 Intron XP_011521026.1
XM_011522727.2 Intron XP_011521029.1
XM_017023819.1 Intron XP_016879308.1
XM_017023820.1 Intron XP_016879309.1
XM_017023821.1 Intron XP_016879310.1

View Full Product Details