Product Details
- SNP ID
-
rs1046306
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:372131 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CGCTCCCGGCCAGCAGGATGTGCCA[A/G]ATGCTGTGGGTGTAGTAGTAGTTGT
- Phenotype
-
MIM: 604853
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
MRPL28
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs113865847] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- MRPL28
- Gene Name
- mitochondrial ribosomal protein L28
There are no transcripts associated with this gene.
- Gene
- TMEM8A
- Gene Name
- transmembrane protein 8A
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_021259.2 |
2301 |
Silent Mutation |
ATC,ATT |
I724I |
NP_067082.2 |
View Full Product Details