Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001164.4 | Intron | NP_001155.1 | |||
NM_001257319.2 | Intron | NP_001244248.1 | |||
NM_001257320.2 | Intron | NP_001244249.1 | |||
NM_001257321.2 | Intron | NP_001244250.1 | |||
NM_001257323.2 | Intron | NP_001244252.1 | |||
NM_001257325.2 | Intron | NP_001244254.1 | |||
NM_001257326.2 | Intron | NP_001244255.1 | |||
NM_145689.2 | Intron | NP_663722.1 | |||
XM_006718209.3 | Intron | XP_006718272.2 | |||
XM_006718210.3 | Intron | XP_006718273.2 | |||
XM_006718211.1 | Intron | XP_006718274.1 | |||
XM_006718212.3 | Intron | XP_006718275.2 | |||
XM_006718213.1 | Intron | XP_006718276.1 | |||
XM_006718214.3 | Intron | XP_006718277.2 | |||
XM_006718215.3 | Intron | XP_006718278.3 | |||
XM_011520037.2 | Intron | XP_011518339.1 | |||
XM_011520038.2 | Intron | XP_011518340.1 | |||
XM_011520040.2 | Intron | XP_011518342.1 | |||
XM_011520042.1 | Intron | XP_011518344.1 | |||
XM_017017639.1 | Intron | XP_016873128.1 | |||
XM_017017640.1 | Intron | XP_016873129.1 | |||
XM_017017641.1 | Intron | XP_016873130.1 | |||
XM_017017642.1 | Intron | XP_016873131.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_000543.4 | Intron | NP_000534.3 | |||
NM_001007593.2 | Intron | NP_001007594.2 | |||
NM_001318087.1 | Intron | NP_001305016.1 | |||
NM_001318088.1 | Intron | NP_001305017.1 | |||
XM_011520303.1 | Intron | XP_011518605.1 | |||
XM_011520304.1 | Intron | XP_011518606.1 |