Product Details

SNP ID
rs4346023
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:111847738 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCCATCATCACACTCTGGTGTATAA[C/T]ATTTCCATCCCTGCTAGAAAGTTCC
Phenotype
MIM: 606723
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MAPKAPK5 PubMed Links

Gene Details

Gene
MAPKAPK5
Gene Name
mitogen-activated protein kinase-activated protein kinase 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003668.3 Intron NP_003659.2
NM_139078.2 Intron NP_620777.1
XM_017020125.1 Intron XP_016875614.1
XM_017020126.1 Intron XP_016875615.1
XM_017020127.1 Intron XP_016875616.1
XM_017020128.1 Intron XP_016875617.1
XM_017020129.1 Intron XP_016875618.1
XM_017020130.1 Intron XP_016875619.1
XM_017020131.1 Intron XP_016875620.1
XM_017020132.1 Intron XP_016875621.1
XM_017020133.1 Intron XP_016875622.1
XM_017020134.1 Intron XP_016875623.1
XM_017020135.1 Intron XP_016875624.1
XM_017020136.1 Intron XP_016875625.1
XM_017020137.1 Intron XP_016875626.1
XM_017020138.1 Intron XP_016875627.1
XM_017020139.1 Intron XP_016875628.1
Gene
MAPKAPK5-AS1
Gene Name
MAPKAPK5 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details