Product Details

SNP ID
rs28419182
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:177301399 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTGAGGCCAGCCGGGTGGCCCCAG[G/A]AAGAACTAGGTGTCTTGACACCTAA
Phenotype
MIM: 609450 MIM: 606681 MIM: 605733 MIM: 612415
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
MXD3 PubMed Links

Gene Details

Gene
MXD3
Gene Name
MAX dimerization protein 3
There are no transcripts associated with this gene.

Gene
NSD1
Gene Name
nuclear receptor binding SET domain protein 1
There are no transcripts associated with this gene.

Gene
PRELID1
Gene Name
PRELI domain containing 1
There are no transcripts associated with this gene.

Gene
RAB24
Gene Name
RAB24, member RAS oncogene family
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001031677.3 1326 UTR 3 NP_001026847.1
NM_130781.3 1326 UTR 3 NP_570137.2

View Full Product Details