Product Details

SNP ID
rs35122441
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:99434723 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTAGTATTTACAAGAAGTAGAAAA[A/C]GTATATATTTTTCTGTTAAGAATAT
Phenotype
MIM: 616653
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
LOC101927365 PubMed Links

Gene Details

Gene
LOC101927365
Gene Name
uncharacterized LOC101927365
There are no transcripts associated with this gene.

Gene
PNISR
Gene Name
PNN interacting serine and arginine rich protein
There are no transcripts associated with this gene.

Gene
TSTD3
Gene Name
thiosulfate sulfurtransferase (rhodanese)-like domain containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001195131.1 2854 Intron NP_001182060.1
XM_017010141.1 2854 Intron XP_016865630.1
Gene
USP45
Gene Name
ubiquitin specific peptidase 45
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080481.1 2854 UTR 3 NP_001073950.1
XM_005267169.3 2854 UTR 3 XP_005267226.1
XM_005267170.4 2854 UTR 3 XP_005267227.1
XM_005267172.3 2854 UTR 3 XP_005267229.1
XM_005267175.3 2854 Intron XP_005267232.1
XM_011536200.2 2854 UTR 3 XP_011534502.1
XM_017011381.1 2854 UTR 3 XP_016866870.1
XM_017011382.1 2854 UTR 3 XP_016866871.1
XM_017011383.1 2854 UTR 3 XP_016866872.1
XM_017011384.1 2854 UTR 3 XP_016866873.1
XM_017011385.1 2854 Intron XP_016866874.1
XM_017011386.1 2854 Intron XP_016866875.1

View Full Product Details