Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001287059.1 | 227 | UTR 5 | NP_001273988.1 | ||
NM_015052.4 | 227 | UTR 5 | NP_055867.3 | ||
XM_005249665.3 | 227 | UTR 5 | XP_005249722.1 | ||
XM_006715670.3 | 227 | UTR 5 | XP_006715733.1 | ||
XM_006715671.3 | 227 | UTR 5 | XP_006715734.1 | ||
XM_006715673.3 | 227 | UTR 5 | XP_006715736.1 | ||
XM_011515220.2 | 227 | Intron | XP_011513522.1 | ||
XM_011515222.2 | 227 | Intron | XP_011513524.1 | ||
XM_011515223.2 | 227 | Intron | XP_011513525.1 | ||
XM_011515224.2 | 227 | Intron | XP_011513526.1 | ||
XM_011515225.2 | 227 | Intron | XP_011513527.2 | ||
XM_011515226.2 | 227 | Intron | XP_011513528.1 | ||
XM_017011882.1 | 227 | Intron | XP_016867371.1 | ||
XM_017011883.1 | 227 | UTR 5 | XP_016867372.1 | ||
XM_017011884.1 | 227 | Intron | XP_016867373.1 | ||
XM_017011885.1 | 227 | UTR 5 | XP_016867374.1 | ||
XM_017011886.1 | 227 | UTR 5 | XP_016867375.1 | ||
XM_017011887.1 | 227 | Intron | XP_016867376.1 | ||
XM_017011888.1 | 227 | Intron | XP_016867377.1 | ||
XM_017011889.1 | 227 | Intron | XP_016867378.1 | ||
XM_017011890.1 | 227 | UTR 5 | XP_016867379.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
XM_017012919.1 | 227 | Intron | XP_016868408.1 |