Product Details

SNP ID
rs2236939
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:50206077 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TAGAGCCCGCGCAGCGCACGGCGGA[G/T]AGAAGGAACCAAGACTTGGTTTGCA
Phenotype
MIM: 139330 MIM: 604437
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
GNAT1 PubMed Links

Gene Details

Gene
GNAT1
Gene Name
G protein subunit alpha transducin 1
There are no transcripts associated with this gene.

Gene
SLC38A3
Gene Name
solute carrier family 38 member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006841.5 Intron NP_006832.1
XM_006712954.2 Intron XP_006713017.1

View Full Product Details