Product Details

SNP ID
rs3810936
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.9:114790605 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
GGAACCAGTTGCTACCTACTTCGCA[C/T]ACAGACTTGGTCCCCATGAGGAGCT
Phenotype
MIM: 604052
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TNFSF15 PubMed Links

Gene Details

Gene
TNFSF15
Gene Name
tumor necrosis factor superfamily member 15
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001204344.1 719 Silent Mutation GTA,GTG V142V NP_001191273.1
NM_005118.3 719 Silent Mutation GTA,GTG V201V NP_005109.2

View Full Product Details