Product Details

SNP ID
rs10114470
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:114785492 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTCAATGAACAAAAGGCCACATAAT[C/T]ATAGATTTGAAAAAGACCTCAGAGA
Phenotype
MIM: 604052
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TNFSF15 PubMed Links

Gene Details

Gene
TNFSF15
Gene Name
tumor necrosis factor superfamily member 15
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001204344.1 5832 UTR 3 NP_001191273.1
NM_005118.3 5832 UTR 3 NP_005109.2

View Full Product Details