Product Details

SNP ID
rs4072865
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.16:10930248 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CCCAGCTCACTATCCTTTCCGCAAA[C/T]TTCACTGAACATCTCGCAGAACTTG
Phenotype
MIM: 600005
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CIITA PubMed Links

Gene Details

Gene
CIITA
Gene Name
class II, major histocompatibility complex, transactivator
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000246.3 Intron NP_000237.2
NM_001286402.1 Intron NP_001273331.1
NM_001286403.1 Intron NP_001273332.1
XM_006720880.3 Intron XP_006720943.2
XM_011522484.2 Intron XP_011520786.1
XM_011522485.2 Intron XP_011520787.1
XM_011522486.2 Intron XP_011520788.1
XM_011522487.2 Intron XP_011520789.1
XM_011522488.2 Intron XP_011520790.1
XM_011522489.2 Intron XP_011520791.1
XM_011522490.2 Intron XP_011520792.1
XM_011522491.2 Intron XP_011520793.1
XM_011522492.2 Intron XP_011520794.1
XM_011522493.2 Intron XP_011520795.1
XM_011522494.2 Intron XP_011520796.1
XM_011522495.2 Intron XP_011520797.1
XM_011522496.2 Intron XP_011520798.1
Gene
DEXI
Gene Name
Dexi homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014015.3 Intron NP_054734.2
XM_011522455.2 Intron XP_011520757.1
XM_017023171.1 Intron XP_016878660.1

View Full Product Details