Product Details

SNP ID
rs6755690
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.2:202225870 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CATAACCATTTGTTTGCATATCTGT[C/G]TCTCCTATCAGATTTGAGTTTCTTA
Phenotype
MIM: 601912
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
SUMO1 PubMed Links

Gene Details

Gene
SUMO1
Gene Name
small ubiquitin-like modifier 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005781.1 Intron NP_001005781.1
NM_001005782.1 Intron NP_001005782.1
NM_003352.4 Intron NP_003343.1

View Full Product Details